Learn More
Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot 1:2000 - 1:10000, ELISA 1:70000, Immunohistochemistry 1:500-1:2500 |
| Host Species | Donkey |
| Immunogen | Donkey anti-Mouse IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Mouse IgG whole molecule in donkey |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
