Learn More
Donkey anti-Mouse IgG (H+L) Secondary Antibody, PerCP, R&D Systems™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Formulation | PBS |
| Host Species | Donkey |
| Immunogen | Mouse IgG whole molecule |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.