Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey Anti-Mouse IgG NorthernLights™ NL637-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. NL008 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL008 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL008 Supplier R&D Systems Supplier No. NL008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody has been used in 9 publications

IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.

Specifications

Antigen IgG
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 1mg/mL
Conjugate NorthernLights 637 (NL637)
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.