Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey Anti-Rabbit IgG Antibody (H&L) Antibody (FITC), ProSci
SDP

Catalog No. 16838655
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-838-655 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-838-655 Supplier ProSci LLC Supplier No. XG6157F0500
Add to Cart
Add to Cart

Secondary Antibody

Donkey Anti-Rabbit IgG Antibody specifically detects Bovine Anti-Human IgG in Rabbit Samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.

Specifications

Antigen Donkey Anti-Rabbit IgG
Applications ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate FITC
Dilution This antibody can be used at 1:100-1:2000 for most IHC/ICC applications. Customers need to optimize the dilutions in their system.
Formulation Buffer containing 0.01M sodium phosphate, 0.25M sodium chloride, pH 7.6 and 25% glycerol.
Host Species Donkey
Immunogen Purified rabbit IgG.
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This conjugate specifically reacts with rabbit IgG with minimal cross-reaction to IgG of other mammalian species.
Target Species Rabbit
Content And Storage Store 2 - 8 °C as an undiluted liquid for several weeks. After dilution, do not use for more than one day. -20 °C for up to one year.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.