Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Donkey anti-Rat IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ Plus 647

Donkey Polyclonal Secondary Antibody

$186.00 - $408.00

Specifications

Antigen Rat IgG (H+L) Highly Cross-Adsorbed
Concentration 2 mg/mL
Content And Storage 4°C, store in dark
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A48272
Encompass_Preferred
View Documents
Invitrogen™
A48272
1 mg
Each for $408.00
Only null left
Add to Cart
 
A48272TR
Encompass_Preferred
View Documents
Invitrogen™
A48272TR
100 μg
Each for $186.00
Only null left
Edge
Add to Cart
 
Description

Description

To minimize cross-reactivity, the donkey anti-rat IgG whole antibodies have been cross-adsorbed against serum proteins from bovine, goat, rabbit, mouse, chicken, guinea pig, hamster, horse, sheep, and human. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in less background staining and cross-reactivity. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. Further passages through additional columns result in highly cross-adsorbed preparations of secondary antibody. The benefits of these extra steps are apparent in multiplexing/multicolor-staining experiments where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins. Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. Specificity: This antibody binds to heavy chains o...

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Rat IgG (H+L) Highly Cross-Adsorbed
4°C, store in dark
Polyclonal
Liquid
IgG
Rat
Gamma Immunoglobins Heavy and Light chains.
Secondary Antibody
2 mg/mL
Western Blot, Immunocytochemistry
Alexa Fluor Plus 647
Donkey
RUO
proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5
Secondary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.