Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP175377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175377 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175377 Supplier Novus Biologicals Supplier No. NBP175377
Add to Cart
Add to Cart

Donkey Polyclonal Antibody has been used in 3 publications

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 4.5mg/mL
Conjugate Unconjugated
Dilution Immunoassay
Host Species Donkey
Immunogen Purified Rat IgG (H&L)
Purification Method Affinity purified
Quantity 2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on rat IgG and light chains on all rat immunoglobulins. This antibody has been pre-adsorbed against bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rabbit and sheep IgG.
Target Species Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.