Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey Anti-Sheep IgG (H+L) Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. F0125 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0125 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F0125 Supplier R&D Systems Supplier No. F0125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Sheep samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG
Applications Flow Cytometry
Classification Polyclonal
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Immunogen Sheep IgG
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Sheep IgG specific IgG was purified by sheep IgG (H+L) affinity chromatography and absorbed against human IgG, and human, mouse, bovine, and rabbit serum to eliminate cross-reactivity.
Target Species Sheep
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.