Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NBP173267 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173267 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173267 Supplier R&D Systems Supplier No. NBP173267

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications Dot Blot, ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:3000 - 1:15000, ELISA 1:50000 - 1:150000, Immunohistochemistry 1:1000 - 1:5000, Dot Blot
Host Species Donkey
Immunogen Donkey anti-Sheep IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Sheep IgG whole molecule in donkey.
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This product was prepared from monospecific antiserum by immunoaffinity chromatography using Sheep IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Donkey Serum, Sheep IgG and Sheep Serum.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.