Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP173720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173720 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173720 Supplier Novus Biologicals Supplier No. NBP173720
Add to Cart
Add to Cart

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:1000 - 1:10000, ELISA 1:65000, Immunohistochemistry 1:500 - 1:2500
Host Species Donkey
Immunogen Donkey anti-Sheep IgG (H+L) Secondary Antibody [HRP] (Pre-adsorbed) was produced by repeated immunization with Sheep IgG whole molecule in donkey.
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This antibody was pre-adsorbed against Chicken, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit, and Rat Serum Proteins. No reaction was observed against Chicken, Guinea Pig, Hamster, Horse, Human, Mouse, Rabbit and Rat Serum Proteins.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.