Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB483971 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB483971 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB483971 Supplier R&D Systems Supplier No. NBP172745PCP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for ELISA, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Host Species Donkey
Immunogen Donkey anti-Sheep IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with Sheep IgG whole molecule in donkey.
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.