Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Sheep IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP175433 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175433 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175433 Supplier R&D Systems Supplier No. NBP175433

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Sheep samples. It is validated for Chemiluminescence Immunoassay, ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot.

Specifications

Antigen IgG (H+L)
Applications Chemiluminescence Immunoassay (CIA), ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot
Classification Polyclonal
Concentration 4.5mg/mL
Conjugate Unconjugated
Dilution Immunoassay
Host Species Donkey
Immunogen Purified Sheep IgG (H&L)
Purification Method Affinity purified
Quantity 2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on sheep IgG and light chains on all sheep immunoglobulins. This antibody has been pre-adsorbed against human and rabbit IgG.
Target Species Sheep
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.