Learn More
Donkey anti-Sheep IgG (H+L) Secondary Antibody, Texas Red (Pre-adsorbed), R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Texas Red |
| Dilution | Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 |
| Host Species | Donkey |
| Immunogen | Sheep IgG whole molecule |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.