Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NB7581 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7581 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7581 Supplier Novus Biologicals Supplier No. NB7581
Add to Cart
Add to Cart

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate PE
Dilution Western Blot 1:1000 - 10000, Flow Cytometry 1:100 - 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100 - 1:200
Host Species Donkey
Immunogen Donkey F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with Rabbit IgG whole molecule in donkey.
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Donkey IgG F(c) or to Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat & Sheep Serum Proteins.
Reconstitution Reconstitute to desired concentration.
Target Species Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.