Learn More
Donkey F(ab')2 anti-Sheep IgG (H+L) Secondary Antibody, PE (Pre-adsorbed), R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | PE |
| Dilution | Flow Cytometry 1:100-1:250, Immunocytochemistry/Immunofluorescence 1:100-1:250 |
| Host Species | Donkey |
| Immunogen | Donkey F(ab')2 anti-Sheep IgG (H+L) Secondary Antibody [PE] (Pre-adsorbed) was produced by repeated immunization with sheep IgG whole molecule in donkey. |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.