Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dopamine beta Hydroxylase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534664 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534664 Supplier Invitrogen™ Supplier No. PA534664
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: SH-SY5Y. Predicted reactivity: Rhesus Monkey (86%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is an oxidoreductase belonging to the copper type II, ascorbate-dependent monooxygenase family. It is present in the synaptic vesicles of postganglionic sympathetic neurons and converts dopamine to norepinephrine. It exists in both soluble and membrane-bound forms, depending on the absence or presence, respectively, of a signal peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dopamine beta Hydroxylase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene DBH
Gene Accession No. P09172, Q05754, Q64237
Gene Alias Dbh; DBM; dopamine beta hydroxylase; Dopamine beta hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase; dopamine beta-hydroxylase (dopamine beta-monooxygenase); dopamine beta-hydroxylase precursor; dopamine beta-hydroxylase precursor (EC 1.14.17.1); dopamine beta-monooxygenase; dopamine beta-monooxygenase precursor (EC 1.14.17.1); Dopamine-Beta-hydroxylase; DOPBHY; mixed function oxidase; Soluble dopamine beta-hydroxylase
Gene Symbols DBH
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Dopamine beta Hydroxylase. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13166, 1621, 25699
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.