Learn More
U.S. Pharmacopeia Dopamine hydrochloride, 62-31-7, MFCD00012898, 200 mg
Supplier: U.S. Pharmacopeia 1225204
Linear Formula: (HO)2C6H3CH2CH2NH2·HCl, Molecular Weight: 189.64, mp: 248-250 °C, Synonym: 2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride.