Learn More
Invitrogen™ Dopamine Transporter Polyclonal Antibody

Description
Immunogen sequence: MSKSKCSVGL MSSVVAPAKE PNAVGPKEVE LILVKEQNGV QLTSSTLTNP RQSPVEAQDR ETWGKKIDFL LSVIGFAVDL ANVWRFPYLC YKNGGGAFLV.
Specifications
Specifications
| Antigen | Dopamine Transporter |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.84 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | Slc6a3 |
| Gene Accession No. | P23977, Q01959, Q61327 |
| Gene Alias | DA transporter; Dat; Dat1; dopamine transporter; dopamine transporter 1; PKDYS; SLC6A3; Sodium-dependent dopamine transporter; solute carrier family 6 (neurotransmitter transporter), member 3; solute carrier family 6 (neurotransmitter transporter, dopamine), member 3; solute carrier family 6 member 3; solute carrier family 6, member 3 |
| Gene Symbols | Slc6a3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.