Learn More
Accuform Signs DOT Shipping Labels
Catalog No. 19804962 Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Corrosive 8
Flammable Gas 2
Flammable Liquid 3
Material:
Paper
Vinyl
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text Material
19-804-962 Flammable Liquid 3 Vinyl
19-809-835 Flammable Gas 2 Paper
19-809-837 Corrosive 8 Paper
19-809-836 Flammable Liquid 3 Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-804-962 Supplier Accuform Signs Supplier No. MSL301EV2
Add to Cart
Add to Cart
to place proper identification and classification of hazardous material during transporting and shipping
Specifications
Adhesive Type Self-adhesive
Format Label
Material Vinyl
Product Type DOT Shipping Label
Display Text Flammable Liquid 3
For Use With (Application) The durable adhesive will stick to most flat, clean surfaces, Adhesive Vinyl signs will work for short term outdoor applications and in moderate temperature
Life Span 2 to 4 Years
Quantity 250/Pk.
Thickness (English) 4 mil
Height (English) 4 in.
Height (Metric) 10.16 cm