Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs DOT Shipping Labels

Catalog No. 19809836 Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Corrosive 8
Flammable Gas 2
Flammable Liquid 3
Material:
Paper
Vinyl
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text Material
19-809-836 Flammable Liquid 3 Paper
19-804-962 Flammable Liquid 3 Vinyl
19-809-835 Flammable Gas 2 Paper
19-809-837 Corrosive 8 Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-809-836 Supplier Accuform Signs Supplier No. MSL301PS2
Add to Cart
Add to Cart

to place proper identification and classification of hazardous material during transporting and shipping

Specifications

Adhesive Type Self-adhesive
Format Label
Material Paper
Product Type DOT Shipping Label
Display Text Flammable Liquid 3
For Use With (Application) Shipping
Includes Graphic
Quantity 250/Pk.
Width (English) 4 in.
Height (English) 4 in.
Height (Metric) 10.16 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.