Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: EXPLOSIVE 1.2D
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
$50.75
Specifications
Color | Black, Orange |
---|---|
Product Type | Vehicle Placard |
Display Text | EXPLOSIVE 1.2D |
For Use With (Application) | Material Handling, Shipping & Receiving Signs & Labels |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-111-274
|
Brady™
63346 |
Fiberglass |
Each for $50.75
|
|
|||||
19-111-196
|
Brady™
63311 |
Pressure Sensitive Vinyl | N/A | N/A | N/A | ||||
19-111-305
|
Brady™
63381 |
Polycoated Tagstock | N/A | N/A | N/A | ||||
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
Black, Orange | |
EXPLOSIVE 1.2D | |
1 Ea. | |
10.75 x 10.75 in. |
Vehicle Placard | |
Material Handling, Shipping & Receiving Signs & Labels | |
Diamond |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title