Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.4G

Catalog No. 19111192 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass
Polycoated Tagstock
Pressure-sensitive Vinyl
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-111-192 Pressure-sensitive Vinyl
19-111-270 Fiberglass
19-111-301 Polycoated Tagstock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-111-192 Supplier Brady™ Supplier No. 63306
Add to Cart
Add to Cart

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Orange
Material Pressure-sensitive Vinyl
Product Type Vehicle Placard
Display Text EXPLOSIVE 1.1G
For Use With (Application) Material Handling, Shipping & Receiving Signs & Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.004 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.