Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: (Flammable Picto w/Blank Box) 3

Catalog No. 19110998 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass (B-120)
Pressure-sensitive Vinyl (B-946)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-110-998 Pressure-sensitive Vinyl (B-946)
19-111-166 Fiberglass (B-120)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-110-998 Supplier Brady™ Supplier No. 60379

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

  • Available in two different materials Pressure Sensitive Vinyl and Premium Fiberglass

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color White, Red
Material Pressure-sensitive Vinyl (B-946)
Product Type Vehicle Placard
Display Text Flammable pictogram with blank box, 3
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.004 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.