Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: (Flammable picto w/ blank box and red/white stripes) 4
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Specifications
Color | Black, Red, White |
---|---|
Product Type | Vehicle Placard |
Display Text | Flammable pictogram with blank box and red/white stripes, 4 |
For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-110-999
|
Brady™
60380 |
Pressure Sensitive Vinyl (B-946) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-111-167
|
Brady™
60412 |
Fiberglass (B-120) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Available in two different materials Pressure Sensitive Vinyl, Premium Fiberglass
Specifications
Black, Red, White | |
Flammable pictogram with blank box and red/white stripes, 4 | |
1 Ea. | |
10.75 x 10.75 in. |
Vehicle Placard | |
Material Handling, Shipping and Receiving Signs and Labels | |
Diamond |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title