Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: FLAMMABLE SOLID 4

Catalog No. 19111334 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass (B-120)
Polycoated Tagstock (B-101)
Pressure-sensitive Vinyl (B-946)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-111-334 Fiberglass (B-120)
19-111-326 Pressure-sensitive Vinyl (B-946)
19-111-349 Polycoated Tagstock (B-101)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-111-334 Supplier Brady™ Supplier No. 63425
Add to Cart
Add to Cart

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass

Specifications

Adhesive Type Non-adhesive
Color Black, Red, White
Material Fiberglass (B-120)
Product Type Vehicle Placard
Display Text FLAMMABLE SOLID 4
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.0984 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.