Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: INHALATION HAZARD 6

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

$27.25

Specifications

Color Black, White
Product Type Vehicle Placard
Display Text INHALATION HAZARD 6
For Use With (Application) Material Handling, Shipping & Receiving Signs & Labels
Quantity 1 Ea.
View More Specs

Products 3
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-111-329
missing translation for 'viewDocuments'
Brady™
63418
Pressure-sensitive Vinyl (B-946)
Each for $27.25
Only null left
Add to cart
 
19-111-338
missing translation for 'viewDocuments'
Brady™
63431
Fiberglass (B-120) N/A N/A N/A
19-111-354
missing translation for 'viewDocuments'
Brady™
63452
Polycoated Tagstock (B-101) N/A N/A N/A
Description

Description

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications

Specifications

Black, White
INHALATION HAZARD 6
1 Ea.
10.75 x 10.75 in.
Vehicle Placard
Material Handling, Shipping & Receiving Signs & Labels
Diamond
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.