Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ DOT Vehicle Placards: OXIDIZER 5.1

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

$6.80 - $53.25

Specifications

Color White, Blue
Product Type Vehicle Placard
Display Text OXIDIZER 5.1
For Use With (Application) Material Handling, Shipping & Receiving Signs & Labels
Quantity 1 Ea.
View More Specs

Products 3
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-111-336
View Documents
Brady™
63428
Fiberglass (B-120)
Each for $53.25
Only null left
Add to Cart
 
19-111-352
View Documents
Brady™
63449
Polycoated Tagstock (B-101)
Each for $6.80
Only null left
Add to Cart
 
19-111-328
View Documents
Brady™
63415
Pressure Sensitive Vinyl (B-946) N/A N/A N/A
Description

Description

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications

Specifications

White, Blue
OXIDIZER 5.1
1 Ea.
10.75 x 10.75 in.
Vehicle Placard
Material Handling, Shipping & Receiving Signs & Labels
Diamond
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.