Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ DOT Vehicle Placards: POISON GAS 2
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
$27.00
Specifications
Color | Black, White |
---|---|
Product Type | Vehicle Placard |
Display Text | POISON GAS 2 |
For Use With (Application) | Material Handling, Shipping and Receiving Signs and Labels |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
19-111-178
|
Brady™
60433 |
Pressure-sensitive Vinyl (B-946) |
Each for $27.00
|
|
|||||
19-111-172
|
Brady™
60423 |
Fiberglass (B-120) | N/A | N/A | N/A | ||||
19-111-176
|
Brady™
60428 |
Polycoated Tagstock (B-101) | N/A | N/A | N/A | ||||
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
Black, White | |
POISON GAS 2 | |
1 Ea. | |
10.75 x 10.75 in. |
Vehicle Placard | |
Material Handling, Shipping and Receiving Signs and Labels | |
Diamond |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title