Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOT1L Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188509
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2996 10 μL
50-199-2995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2996 Supplier Active Motif Supplier No. 39954
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone methyltransferase. Methylates 'Lys-79' of histone H3. Nucleosomes are preferred as substrate compared to free histones. Binds to DNA.

Specifications

Antigen DOT1L
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene Dot1l
Gene Accession No. Q8TEK3
Gene Alias A630076O07; AW907654; DKFZp586P1823; DOT1; DOT1 like histone H3K79 methyltransferase; DOT1 like histone lysine methyltransferase; DOT1L; DOT1-like histone H3 methyltransferase; DOT1-like histone H3K79 methyltransferase; DOT1-like protein; DOT1-like, histone H3 methyltransferase; DOT1-like, histone H3 methyltransferase (S. cerevisiae); H3-K79-HMTase; histone H3 methyltransferase DOT1; Histone H3-K79 methyltransferase; histone methyltransferase DOT1L; Histone-lysine N-methyltransferase, H3 lysine-79 specific; KIAA1814; KMT4; Lysine N-methyltransferase 4; mDot1
Gene Symbols Dot1l
Host Species Rabbit
Immunogen This DOT1L antibody was raised against a peptide derived from the C-terminal region of human DOT1L.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84444
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.