Learn More
CPC Scientific 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-Thr-OH
(Disulfide bond) 5MG
SDP
Supplier:  CPC Scientific SOMA011B
Encompass
DOTA-conjugated somatostatin analog for labeling with radiometals such as gallium-67 or gallium-68, indiuM-111, and yttrium-90. Potentially useful for receptor-mediated tumor imaging and peptide receptor-targeted radionuclide therapy.SEQUENCE: 4,7,10-Tricarboxymethyl-1,4,7,10-tetraaza-cyclododecan-1-yl-acetyl-D-Phe-Cys-Tyr-D-Trp-Lys-Thr-Cys-Thr-OH(Disulfide bond)ONE-LETTER SEQUENCE: DOTA-fCYwKTCT (Cys & Cys bridge)MOLECULAR FORMULA: C65H90N14O19S2MOLECULAR WEIGHT: 1435.65STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [177943-89-4]SYNONYMS: DOTA-(Tyr)-Octreotate, DOTA-[Tyr3] Octreotide Acid (Octreotate), DOTATATE, DOTA-(Tyr,Thr)-Octreotide, DOTATATERESEARCH AREA: GastrointestinalREFERENCES:W.A.Breeman et al., Eur. J. Nucl. Med. Mol. Imaging, 30, 917 (2003)A.Capello et al., Cancer Biother. Radiopharm., 18, 761 (2003)M.de Jong et al., J. Nucl. Med., 46, 13S (2005)L.Kolby et al., Br. J. Cancer, 93, 1144 (2005)R.Valkema et al., J. Nucl. Med., 4
Catalog No. 50-211-0438
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.