Learn More
Medchemexpress LLC HY-112754A 50mg Medchemexpress, DOTAP (chloride) CAS:132172-61-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112754A50MG
Encompass_Preferred
Medchemexpress, HY-112754A 50mg DOTAP (chloride) CAS:132172-61-3 DOTAP chloride is a useful and effective cationic lipid for transient and stable transfection DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides) with out the use of helper lipid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.