Learn More
Poltex Double Bulk Bin With Lid, 4 in. Wide Comp
SDP
Catalog No. 21377540
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Wall Mount or Countertop
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-540 Wall Mount or Countertop
21-377-538 Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-540 Supplier Poltex Supplier No. BLKBNLDDBL4W
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Simple 4 in. two-compartment bulk bin with top opening with lid for filling and front openings for dispensing.
Simple two-compartment bulk bin with lid
Durable and cleanable PETG construction
4 in. wide
Choose from:
  • Wall or countertop mount with rubber bumpons on base
  • VHB tape mount (very high bond tape included)
  • Magnet mount (powerful magnets included)
Specifications
Type Dispensing Bin
Material PETG
Color Clear
Shape Square
Mounting Type Wall Mount or Countertop
Autoclavable Not Autoclavable