Learn More
Description
- High shear adhesive with high temperature resistance
- Durable foam is ideal for interior mounting applications
- Ideal for bonding, attaching, and mounting
- Thick foam dampens and prevents vibration and open-cell structure aids component cooling
- Formulated for hard-to-bond surfaces.
- Unique rubber adhesive system provides quick stick and conforms to irregular and low surface energy (LSE) materials.
Specifications
Specifications
| Product Type | Foam Tape |
| For Use With (Application) | Appliance, Bonding and Assembly, Building Maintenance and Repair, Defense, Heavy Industrial, Marine, Metalworking, Personal Recreational Vehicles, Rail |
| Adhesive Type | Rubber |
| Color | White |
| Dimensions (L x W) | 36 yd. x 1 in. |
| Material | Polyethylene Foam |
| Model | 4466 |
| Quantity | 9 Rolls/Cs. |
| Thickness (English) | 62 mil |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.