Learn More
SPECTRUM SCIENTIFICS DOUBLE CONCAVE SLIDES, 72/PK
Supplier: SPECTRUM SCIENTIFICS 5035
Double Concavity Slides are 1.2-1.3mm thick with two 15mm diameter wells approximately. 0.5mm deep. Packed is boxes of 12 slides per box. Double Concave slides have two 'caves' for side-by-side sample viewing. 72 slides per pack.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.