Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Double Convex Acrylic Lenses
SCI_ED

Catalog No. S27416 Shop All Eisco Products
Change view
Click to view available options
Length (Metric) Focal:
200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Focal
S27416 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27416 Supplier Eisco™ Supplier No. PH0577A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Double convex acrylic lenses are a great tool for studying optics and the properties of light.

  • One face is clear, the other is frosted
  • Made from high quality acrylic\
  • Length: 89 mm

Specifications

Material Acrylic
Includes One Clear Side, One Frosted Side
For Use With (Application) Studying Optics and Properties of Light
Length (Metric) Focal 200 mm
Product Type Double Concave Lens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.