Learn More
United Scientific Double Convex Lens
SDPSCI_ED
Catalog No. S81063 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Optics Experiments
Catalog No. For Use With (Application)
S81063 Optics Experiments
1 options
Catalog No. S81063 Supplier United Scientific Supplier No. LCV707
Add to Cart
Add to Cart
Double Convex Lens is high quality and safe for students.
  • Made from plate glass
  • Carefully ground and polished optical surfaces, and cleanly ground edges
  • Prop 65 Warning Label
Specifications
For Use With (Application) Optics Experiments
Product Type Lens
Dimensions (L x W) 75 mm dia./175 mm FL