Learn More
Poltex Double Gown Dispenser, Wall Mount
SDP
Catalog No. 22022568
Change view
Click to view available options
Bulk Apparel Dispenser
Catalog No.
22-022-568 Bulk Apparel Dispenser
1 options
Catalog No. 22-022-568 Supplier Poltex Supplier No. GOWN2VRTW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Holds two stacks of isolation gowns or coveralls. Wall mount (screws/anchors included).
  • Holds two stacks of isolation gowns or coveralls
  • Durable and cleanable PETG construction
  • Available in wall mount (screws/anchors included), magnet mount (powerful magnets indcluded), or VHB tape mount (very high bond tape included)
Specifications
Product Type Bulk Apparel Dispenser
Color Clear
Length (Metric) 137.16 mm
Width (Metric) 294.64 mm
Height (Metric) 624.84 mm
Holds Shoe Covers, Bouffant Caps
Material PETG
Mounting Wall Mount
No. of Compartments 2
Product Style Front-loading with Opening
Sterility Non-sterile