Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Double-Layer Full-Face Nomex™ Hood

Catalog No. 19100452 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Black
White
Design Type:
Straight Cut
Tri-Cut
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Design Type
19-100-452 Black Straight Cut
19-100-450 White Tri-Cut
19-100-451 White Straight Cut
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-100-452 Supplier PIP™ Supplier No. 906100NOM7BLKB
  • $119.00
    $94.65 / Each of 1

    Save $24.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PIP™ Double-Layer Full-Face Nomex™ Hood are flame resistant balaclavas that offer limited chemical splash protection along with excellent wear and durability

A key factor in the thermal protection Nomex™ provides is that it carbonizes and thickens when exposed to the intense heat of fires. This increases the protective barrier between the heat source and the skin and minimizes burn injury. This tough, protective barrier stays supple and flexible until it cools, which gives valuable extra seconds of protection when it is most needed.

  • Protective qualities will not wash out
  • Fabric meets OSHA requirements for flame resistance
  • TPP Rating 20-23
  • Meets NFPA 1971 requirements
  • One Size Fits Most

Specifications

Product Type Fire Protectant Hood
Size Universal
Material Nomex
Color Black
Closure Type Pull-over
Disposable Reusable
Design Type Straight Cut
Quantity 1 Ea.
Includes Hood Only
Construction 2-ply head and bib
For Use With (Application) Heat/Fire Applications
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.