Learn More
Berkshire™ Double-Sided Flat Mop Nonwoven Flat Mop Cover
Catalog No. 19070016 Shop All Berkshire Corporation Products
Change view
Click to view available options
Double-Sided Flat Mop Nonwoven Flat Mop Cover; Type: Double-Sided Flat Mop Nonwoven Cover; Material: Nonwoven; Sterility: Non-Sterile; Autoclavable: Autoclavable
Catalog No.
19-070-016 Double-Sided Flat Mop Nonwoven Flat Mop Cover; Type: Double-Sided Flat Mop Nonwoven Cover; Material: Nonwoven; Sterility: Non-Sterile; Autoclavable: Autoclavable
1 options
Catalog No. 19-070-016 Supplier Berkshire™ Supplier No. FMC5NW24
Double-Sided Flat Mop Nonwoven Cover is made of polyester and cellulose with 100% rayon nonwoven inner layers to provide superior absorbency. Designed for the capture and removal of contaminants during the cleaning process. Each cover is individually wrapped to preserve cleanliness.
Double-Sided Flat Mop Nonwoven Cover is made of polyester and cellulose with 100% rayon nonwoven inner layers to provide superior absorbency. Designed for the capture and removal of contaminants during the cleaning process. Each cover is individually wrapped to preserve cleanliness.
  • 68 gsm polyester and cellulose outer cover
  • 100% rayon nonwoven inner layer
  • Individually wrapped to preserve cleanliness
  • Autoclavable
Specifications
Product Type Double-sided Flat Mop Nonwoven Cover
Autoclavable Autoclavable
Color-coded No