Description
- Package includes the replacement Media Bridge only (printer sold separately)
- The media bridge holds two-sided material above the tape roll
- The material lays across the media bridge to enter the tape entrance ramp
- When not in use, the bridge is raised and snaps into the enclosure cover
Specifications
Specifications
| Product Type | Double-sided Printing Adaptor |
| Sterility | Non-sterile |