Learn More
Globe Scientific Single Thumb Tab Snap Caps
Colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
Supplier:  Globe Scientific 11090500
Catalog No. 22-010-004
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
Description
Description
  • Material: Low-density polyethylene (LDPE)
  • Features one thumb tab for easy removal of the cap
  • Allows more tubes to be stacked closely in racks
  • Maintain tight seal during centrifugation, agitation and refrigeration procedures
  • Available in bright attractive colors for sample identification purposes
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.