Learn More
Dowex Marathon C, H+ form, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Chemical Name or Material | Dowex Marathon C |
| Name Note | H+ form |
| CAS | 150604-77-6 |
| MDL Number | MFCD00212649 |
| Density | 1.2 |
| Quantity | 1000 g |
| Solubility Information | Insoluble in water. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.