Learn More
Chempoint Com DOWTHERM SR-1 55 GALLON DRUM
Supplier:  Chempoint Com 0124553
Encompass
Ethylene glycol-based fluid (95%) Fluorescent pink. Efficient heat transfer u-28 to 120 C. Provides freeze / burst protection u-51 C / -60 C (depending on concentration) Dow is the oldest back-integrated supplier of glycol based heat transfer fluids: uDOWTHERM SR1 originated in the late 1940AEs.
Catalog No. NC0318645
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.