Learn More
Doxapram hydrochloride hydrate, Thermo Scientific™
Specifications
Specifications
| Chemical Name or Material | Doxapram hydrochloride hydrate |
| CAS | 7081-53-0 |
| Quantity | 1 g |
| Solubility | Soluble aqueous solution: slightly soluble in chloroform |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C24 H30 N2 O2 . H Cl . x H2 O |
| Physical Form | Crystalline Powder |
| Color | White to Almost-white |
| Purity | ≥98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.