Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Doxazosin mesylate
GREENER_CHOICESDP

Catalog No. 296450
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-645-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-645-0 Supplier Tocris Bioscience™ Supplier No. 2964/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α1 antagonist

Selective α1-adrenoceptor antagonist (pKi values are 9.0, 8.5 and 8.4 for human α1B, α1A and α1D receptors respectively). Displays antihypertensive activity.

Chemical Identifiers

CAS 77883-43-3
Molecular Formula C24H29N5O8S

Specifications

Quantity 50 mg
Formula Weight 547.58
Purity >99%
Molecular Weight (g/mol) 547.583
Chemical Name or Material Doxazosin mesylate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.