Learn More
Doxepin hydrochloride
Description
It is a highly potent H1 histamine receptor antagonist (Kd = 310 pM) and tricyclic antidepressant that inhibits the reuptake of serotonin and norepinephrine and acts as an antagonist at histamine, serotonin, adrenergic, and muscarinic receptors with Ki values in the nanomolar range. Also binds to the H4 histamine receptor (pKi = 6.79). Used clinically to treat anxiety and depression.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 187°C to 189°C |
| Color | White |
| Quantity | 5 g |
| UN Number | UN2811 |
| Merck Index | 14,3435 |
| Solubility Information | Soluble in water (100 mM),chloroform (1:2),alcohol (1:1),methanol,and DMSO. |
| Formula Weight | 315.84 |
| Physical Form | Powder |
| Chemical Name or Material | Doxepin hydrochloride |
Safety and Handling
| Hazard Category | H301-H361 |
| Hazard Statement | GHS H Statement H301-H361 Toxic if swallowed. Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 214-966-8 |
| RTECSNumber | HQ4375000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.