Learn More
Apexbio Technology LLC Gambogic Acid 2752-65-0 10mg
SDP
Supplier:  Apexbio Technology LLC A342410
Encompass_Preferred
Gambogic Acid (CAS 2752-65-0) is a natural cytotoxic compound known to induce apoptosis through interactions with multiple apoptotic regulatory proteins It primarily targets anti-apoptotic Bcl-2 family members including Bcl-XL Bcl-2 Bcl-W Bcl-B Bfl-1 and Mcl-1 competitively binding to their BH3 domains and disrupting their suppression of mitochondrial pro-apoptotic factors It activates caspase-driven apoptotic pathways with EC50 values between 0 78-1 64 M Gambogic Acid demonstrates selective toxicity against cancer cell lines such as human gastric carcinoma MGC-803 cells and U266 multiple myeloma cells and inhibits tumor growth and angiogenesis in various in vivo xenograft models suggesting potential value for cancer biology research
Catalog No. 50-000-64374
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.