Learn More
Selleck Chemical LLC Decitabine S1200-1g
SDP
Supplier:  Selleck Chemical LLC S12001G
Encompass_Preferred
Decitabine is a DNA methyltransferase inhibitor incorporating into DNA and resulting in hypomethylation of DNA and intra-S-phase arrest of DNA replication It is used to treat myelodysplastic syndrome (MDS) Decitabine induces cell cycle arrest and apoptosis in various cancer cell lines
Catalog No. 50-311-1418
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.