Learn More
Cayman Chemical Doxorubicin (hydrochloride), 25316-40-9, 50 mg
SDP
Supplier:  Cayman Chemical 1500750
Encompass_Preferred
Molecular Formula C27H29NO11 • HCl, Purity ≥98%, Formula Weight 580.0, Synonyms: DOX; An anthracycline antitumor antibiotic agent that inhibits DNA topoisomerase II by inducing double-stranded DNA breaks; inhibits nucleic acid synthesis and induces apoptosis by intercalating within DNA.
Catalog No. NC0898359
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.