Learn More
Lkt Laboratories CAS 25316-40-9, ≥98%, DNA intercalator and topoisomerase II inhibitor used to treat various cancers. It inhibits DNA repair by promoting histone H2AX eviction from chromatin.
SDP
Supplier:  Lkt Laboratories D5794
Encompass_Preferred
Doxorubicin is an anthracycline that exhibits anticancer chemotherapeutic and anti-parasitic, antimalarial activities. Doxorubicin is a DNA intercalator that inhibits topoisomerase II and is clinically used to treat various cancers; it is part of the CHOP and ABVD chemotherapy regimens. Doxorubicin also promotes histone H2AX eviction from chromatin, limiting DNA repair mechanisms. Additionally, this compound inhibits growth of Plasmodium.
Catalog No. NC2539794
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.