Learn More
Description
Biological Activity: Antitumor antibiotic agent that inhibits DNA topoisomerase II. DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Reduces intracellular tau levels.
Storage: Store at 4°C.
Note: Inhibitors are guaranteed for 1 year from date of receipt.
Specifications
Specifications
| Color | Orange |
| Product Type | Inhibitor |
| Target Proteins | Doxorubicin hydrochloride |
| CAS | 25316-40-9 |
| Molecular Formula | C30H41FN4O12 |
| Formula Weight | 579.99g/mol |
| Solubility | Soluble to 50nM in DMSO, 50nM water |
| Content And Storage | Store at 4°C |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 10 mg |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
